missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAP2 Recombinant Rabbit Monoclonal Antibody (5I10B3)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558169

 View more versions of this product

Product Code. 30283361

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QPNLPSPGTL APTTLQGVVK FQDVSFAYPN RPDRPVLKGL TFTLRPGEVT ALVGPNGSGK STVAALLQNL YQPTGGQVLL DEKPISQYEH CYLHSQVVSV GQEPVLFSGS VRNNIAYGLQ SCEDDKVMAA AQAAHADDFI QEMEHGIYTD VGEKGSQLAA GQKQRLAIAR ALVRDPRVLI LDEATSALDV QCEQALQDWN SRGDRTVLVI AHRLQTVQRA HQILVLQEGK LQKLAQL.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. This gene is located 7 kb telomeric to gene family member ABCB2. The protein encoded by this gene is involved in antigen presentation. This protein forms a heterodimer with ABCB2 in order to transport peptides from the cytoplasm to the endoplasmic reticulum. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease. Alternative splicing of this gene produces two products which differ in peptide selectivity and level of restoration of surface expression of MHC class I molecules.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TAP2
Recombinant Monoclonal
0.8 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q03519
TAP2
Recombinant fusion protein containing a sequence corresponding to amino acids 450-686 of human TAP2 (NP_001276972.1).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
5I10B3
Unconjugated
TAP2
ABC transporter, MHC 2; ABC18; ABCB3; AI462429; Antigen peptide transporter 2; APT2; ATP dependent transport protein family member; ATP-binding cassette sub-family B member 3; ATP-binding cassette, sub-family B (MDR/TAP), member 3; Cim; D6S217E; Ham2; Ham-2; Histocompatibility antigen modifier 2; jas; MTP2; Peptide supply factor 2; peptide transporter involved in antigen processing 2; Peptide transporter PSF2; Peptide transporter TAP2; PSF2; PSF-2; Really interesting new gene 11 protein; RING11; TAP2; Tap-2; Transporter 2 ABC (ATP binding cassette); transporter 2 ATP-binding cassette sub-family B; transporter 2, ABC (ATP binding cassette); transporter 2, ATP binding cassette subfamily B member; transporter 2, ATP-binding cassette, sub-family B (MDR/TAP); Y1
Rabbit
Protein A
RUO
6891
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.