missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11114-1-AP-150UL
This item is not returnable.
View return policy
Description
Western blot analysis using this antibody detected a major band around 70-80kD in HeLa cells.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.Specifications
| TAP1 | |
| Polyclonal | |
| Unconjugated | |
| TAP1 | |
| ABCB2, Antigen peptide transporter 1, PSF 1, RING 4, RING4, TAP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21354, 24811, 6890 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21958, P36370, Q03518 | |
| TAP1 | |
| TAP1 Fusion Protein Ag1619 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction