missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142408
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.
Specifications
| TAP1 | |
| Polyclonal | |
| Unconjugated | |
| TAP1 | |
| ABC transporter, MHC 1; ABC transporter, MHC, 1; ABC17; ABCB2; ABP-278; ABP-280; ABP-280 homolog; Antigen peptide transporter 1; AOI; APT1; ATP dependent transport protein family member; ATP-binding cassette sub-family B member 2; ATP-binding cassette transporter, major histocompatibility complex, 1; ATP-binding cassette, sub-family B (MDR/TAP), member 2; beta-filamin; Cim; D6S114E; DAAP-57C1.5; FH1; filamin homolog 1; filamin-3; filamin-B; FLJ26666; FLJ41500; FLN1L; FLN-B; Ham1; Ham-1; Histocompatibility antigen modifier 1; LRS1; MTP1; Peptide supply factor 1; peptide transporter involved in antigen processing 1; Peptide transporter PSF1; peptide transporter TAP1; PSF1; PSF-1; Really interesting new gene 4 protein; RING4; SCT; TABP; TAP; tap i; TAP1; Tap-1; TAP1*0102N; TAP1N; Tap2; thyroid autoantigen; transporter 1 ABC (ATP binding cassette); transporter 1 ATP-binding cassette sub-family B; transporter 1, ABC (ATP binding cassette); transporter 1, ATP binding cassette subfamily B member; transporter 1, ATP-binding cassette, sub-family B (MDR/TAP); transporter associated with antigen processing; transporter associated with antigen processing 1; transporter, ABC, MHC, 1; transporter, ATP-binding cassette, major histocompatibility complex, 1; Y3 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 6890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q03518 | |
| TAP1 | |
| Peptide with sequence C-QKFREKLQEIKT (aa 388-399). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction