Learn More
Invitrogen™ TAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110458
Description
Immunogen sequence: IYPRVQKAVG SSEKIFEYLD RTPRCPPSGL LTPLHLEGLV QFQDVSFAYP NRPDVLVLQG LTFTLRPGEV TALVGPNGSG KSTVAALLQN LYQPTGGQLL LDGKPLPQYE HRYLHRQVAA VGQEPQVFGR SLQENIAYGL TQKPTMEEIT AAAVKSGAHS FISGLPQGYD TEVDEAGSQL SGGQRQAVAL ARALIRKPCV LILDDATSAL DANSQLQVEQ LLYESPERYS RSVLLITQHL SLVEQADHIL FLEGGAIREG GTHQQLMEKK GCYWAMVQAP ADAPE.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.
Specifications
| TAP1 | |
| Polyclonal | |
| Unconjugated | |
| TAP1 | |
| ABC transporter, MHC 1; ABC transporter, MHC, 1; ABC17; ABCB2; ABP-278; ABP-280; ABP-280 homolog; Antigen peptide transporter 1; AOI; APT1; ATP dependent transport protein family member; ATP-binding cassette sub-family B member 2; ATP-binding cassette transporter, major histocompatibility complex, 1; ATP-binding cassette, sub-family B (MDR/TAP), member 2; beta-filamin; Cim; D6S114E; DAAP-57C1.5; FH1; filamin homolog 1; filamin-3; filamin-B; FLJ26666; FLJ41500; FLN1L; FLN-B; Ham1; Ham-1; Histocompatibility antigen modifier 1; LRS1; MTP1; Peptide supply factor 1; peptide transporter involved in antigen processing 1; Peptide transporter PSF1; peptide transporter TAP1; PSF1; PSF-1; Really interesting new gene 4 protein; RING4; SCT; TABP; TAP; tap i; TAP1; Tap-1; TAP1*0102N; TAP1N; Tap2; thyroid autoantigen; transporter 1 ABC (ATP binding cassette); transporter 1 ATP-binding cassette sub-family B; transporter 1, ABC (ATP binding cassette); transporter 1, ATP binding cassette subfamily B member; transporter 1, ATP-binding cassette, sub-family B (MDR/TAP); transporter associated with antigen processing; transporter associated with antigen processing 1; transporter, ABC, MHC, 1; transporter, ATP-binding cassette, major histocompatibility complex, 1; Y3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21354, 6890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.76 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21958, Q03518 | |
| TAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 524-808 of human TAP1 (NP_0005843). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.