missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAP1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33388-100ul
This item is not returnable.
View return policy
Description
TAP1 Monoclonal antibody specifically detects TAP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TAP1 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:10000, ELISA Recommended starting concentration is 1 μg/mL | |
| ABC transporter, MHC 1, ABCB2FLJ41500, antigen peptide transporter 1, APT1, ATP-binding cassette sub-family B member 2, ATP-binding cassette, sub-family B (MDR/TAP), member 2, D6S114E, Peptide supply factor 1, Peptide transporter involved in antigen processing 1, Peptide transporter PSF1, Peptide transporter TAP1, PSF-1, PSF1ABC17, Really interesting new gene 4 protein, RING4FLJ26666, TAP1*0102N, TAP1N, transporter 1, ATP-binding cassette, sub-family B (MDR/TAP), transporter associated with antigen processing, transporter, ATP-binding cassette, major histocompatibility complex, 1, Y3 | |
| Recombinant funsion protein containing a sequence corresponding to amino acids 449-748 of human TAP1 (NP_000584.2).,, Sequence:, LYQMQFTQAVEVLLSIYPRVQKAVGSSEKIFEYLDRTPRCPPSGLLTPLHLEGLVQFQDVSFAYPNRPDVLVLQGLTFTLRPGEVTALVGPNGSGKSTVAALLQNLYQPTGGQLLLDGKPLPQYEHRYLHRQVAAVGQEPQVFGRSLQENIAYGLTQKPTMEEITAAAVKSGAHSFISGLPQGYDTEVDEAGSQLSGGQRQAVALARALIRKPCVLILDDATSALDANSQLQVEQLLYESPERYSRSVLLITQHLSLVEQADHILFLEGGAIREGGTHQQLMEKKGCYWAMVQAPADAPE | |
| 100 μL | |
| Cell Biology, Diabetes Research, Membrane Trafficking and Chaperones | |
| 6890 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction