missing translation for 'onlineSavingsMsg'
Learn More

TAP1 Antibody (3D4), Novus Biologicals™

Product Code. 18389379 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389379 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389379 Supplier Novus Biologicals Supplier No. H00006890M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAP1 Monoclonal antibody specifically detects TAP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3D4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14081
Gene Alias ABC transporter, MHC 1, ABCB2FLJ41500, antigen peptide transporter 1, APT1, ATP-binding cassette sub-family B member 2, ATP-binding cassette, sub-family B (MDR/TAP), member 2, D6S114E, Peptide supply factor 1, Peptide transporter involved in antigen processing 1, Peptide transporter PSF1, Peptide transporter TAP1, PSF-1, PSF1ABC17, Really interesting new gene 4 protein, RING4FLJ26666, TAP1*0102N, TAP1N, transporter 1, ATP-binding cassette, sub-family B (MDR/TAP), transporter associated with antigen processing, transporter, ATP-binding cassette, major histocompatibility complex, 1, Y3
Host Species Mouse
Immunogen TAP1 (AAH14081, 241 a.a. ~ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GSETRRLSLFLVLVVLSSLGEMAIPFFTGRLTDWILQDGSADTFTRNLTLMSILTIASAVLEFVGDGIYNNTMGHVHSHLQGEVFGAVLRQETEFFQQNQTGNIMSRVTE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Diabetes Research, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 6890
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.