missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAP Tag Polyclonal Antibody
GREENER_CHOICE

Product Code. 10506450
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10506450 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10506450 Supplier Invitrogen™ Supplier No. CAB1001

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CAB1001 detects TAP. CAB1001 has been successfully used in Western Blot, IP, and ChIP applications The CAB1001 immunogen is a KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage. TAP adds about 20 kDa to the size of the protein.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Tandem affinity purification (TAP) is an affinity purification method for isolation of TAP-tagged proteins together with associated proteins. The protocol involves the fusion of the 'TAP tag' (typically a calmodulin binding peptide (CBP), a tobacco etch virus protease (TEV protease) cleavage site and Protein A) to the protein of interest. The TAP technique is useful in analyzing in vivo interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP Tag
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation BBS with no preservative; pH 8.4
Gene Alias Tobacco Etch Virus
Host Species Rabbit
Immunogen KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.