missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAP Tag Monoclonal Antibody (22F12-1E3), DyLight™ 800 4X PEG
GREENER_CHOICE

Product Code. 15342907
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15342907 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15342907 Supplier Invitrogen™ Supplier No. MA1108D800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-108-D800 detects the Tev cleavage peptide sequence. MA1-108-D800 has been successfully used for fluorescent detection of TAP in Western Blot. DyLight 800 4xPEG has an excitation/emission of 783/797nm.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Tandem affinity purification (TAP) is an affinity purification method for isolation of TAP-tagged proteins together with associated proteins. The protocol involves the fusion of the 'TAP tag' (typically a calmodulin binding peptide (CBP), a tobacco etch virus protease (TEV protease) cleavage site and Protein A) to the protein of interest. The TAP technique is useful in analyzing in vivo interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP Tag
Applications Western Blot
Classification Monoclonal
Clone 22F12-1E3
Concentration 1 mg/mL
Conjugate DyLight 800 4X PEG
Formulation MES with proprietary stabilizer and no preservative
Gene Alias Tobacco Etch Virus
Host Species Mouse
Immunogen KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.