missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521412
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLaS3, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (98%), Rat (97%), Pig (96%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The constitutive transport element (CTE) of type D retroviruses serves as a signal of nuclear export for unspliced viral RNAs. TAP (also known as NXF1) mediates the export of CTE-containing simian type D retroviral RNAs through binding directly to the CTE. TAP is associated with a recognized mRNA export pathway and is a member of the multigene family of NXF proteins. NXF proteins belong to an evolutionarily conserved family of proteins, which are characterized by a leucine-rich-repeat domain (LRR) followed by a region known as the Nuclear Transport Factor 2 (NTF2)-like domain.
Specifications
| TAP | |
| Polyclonal | |
| Unconjugated | |
| NXF1 | |
| Mex 67 homolog; MEX67; Mex67h; modifier of vibrator 1; mRNA export factor TAP; Mvb1; Nuclear RNA export factor 1; nuclear RNA export factor 1 homolog; Nxf1; TAP; tip associated protein; tip associating protein; tip-associated protein; Tip-associating protein; Vdp | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10482, 53319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 10 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99JX7, Q9UBU9 | |
| NXF1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human TAP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction