Learn More
Invitrogen™ Human TAOK2 (TAO1), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic
Specifications
Specifications
| Accession Number | Q9UL54 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 9344 |
| Molecular Weight (g/mol) | 63.1 kDa |
| Name | Human TAOK2 (TAO1), GST Tag |
| Quantity | 10 μg |
| Regulatory Status | For research use only. Not for use in diagnostic procedures |
| Gene Alias | 1110033K02Rik; B230344N16; hKFC-C; Kiaa0881; Kinase from chicken homolog C; MAP3K17; mKIAA0881; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived sterile 20-like kinase 1; PSK; PSK1; PSK-1; PSK1-BETA; serine/threonine protein kinase TAO2; serine/threonine-protein kinase TAO2; serine-threonine kinase; tao; TAO kinase 2; TAO kinase 2 TAO kinase 2; TAO1; TAO2; TAOK2; Thousand and one amino acid protein 2; thousand and one amino acid protein kinase 2; UNQ2971/PRO7431 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.