missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAOK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TAOK2 |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16830815
|
Proteintech
21188-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16820815
|
Proteintech
21188-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The immunogen of this antibody is 742-969 aa of human TAOK2 protein isoform Alpha. This antibody mainly detects the isoforms 1, 3, and 4. It does not detect isoform 2.
In vitro, TAO (thousand and one amino acid) protein kinase 2 (TAO2) activates MAP/ERK kinases (MEK ) 3, 4, and 6 toward their substrates p38 MAP kinase JNK/SAPK (Chen et al., 1999; Chen and Cobb, 2001). This and more recent work has led to the proposal that the TAO protein kinases play an essential role in signaling from physiological agonist to the stress -responsive p38 MAPK (Chen et al., 2003). Autophosphorylation of TAO may play a role in the mechanism of TAO activation. The MEK binding domain of TAO is autophosphorylated on both serine and threonine residues and Ser181 is located within this domain.Specifications
| TAOK2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q6ZQ29, Q9JLS3, Q9UL54 | |
| 381921, 64666, 9344 | |
| TAOK2 Fusion Protein Ag15521 | |
| Primary | |
| -20°C | |
| TAOK2 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hKFC C, KIAA0881, Kinase from chicken homolog C, MAP3K17, PSK, PSK 1, PSK1, TAO kinase 2, TAO1, TAO2, TAOK2 | |
| Taok2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title