missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAOK2 Monoclonal Antibody (2F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520685
This item is not returnable.
View return policy
Description
Peptide Sequence: ELQCRQYKRK MLLARHSLDQ DLLREDLNKK QTQKDLECAL LLRQHEATRE LELRQLQAVQ RTRAELTRLQ HQTELGNQLE YNKRREQELR QKHAAQVRQQ.
TAOK2 (TAO1) is a serine/threonine kinase that may be an important regulator of the stress-responsive p38 MAP kinase pathway. In vitro, TAO (thousand and one amino acid) protein kinase 2 (TAO2) activates MAP/ERK kinases (MEK ) 3, 4, and 6 toward their substrates p38 MAP kinase JNK/SAPK (Chen et al., 1999; Chen and Cobb, 2001). This and more recent work has led to the proposal that the TAO protein kinases play an essential role in signaling from physiological agonist to the stress -responsive p38 MAPK (Chen et al., 2003). Autophosphorylation of TAO may play a role in the mechanism of TAO activation. The MEK binding domain of TAO is autophosphorylated on both serine and threonine residues and Ser181 is located within this domain.
Specifications
| TAOK2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9UL54 | |
| TAOK2 | |
| TAOK2 (AAH51798, 831 a.a. ~ 930 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Proximity Ligation Assay, Immunocytochemistry, Western Blot | |
| 2F4 | |
| Unconjugated | |
| TAOK2 | |
| 1110033K02Rik; B230344N16; hKFC-C; Kiaa0881; Kinase from chicken homolog C; MAP3K17; mKIAA0881; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived sterile 20-like kinase 1; PSK; PSK1; PSK-1; PSK1-BETA; serine/threonine protein kinase TAO2; serine/threonine-protein kinase TAO2; serine-threonine kinase; tao; TAO kinase 2; TAO kinase 2TAO kinase 2; TAO1; TAO2; TAOK2; Thousand and one amino acid protein 2; thousand and one amino acid protein kinase 2; UNQ2971/PRO7431 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9344 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction