missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAOK1 Recombinant Rabbit Monoclonal Antibody (K01_2P66)
Description
TAOK1 Recombinant Monoclonal Antibody for Western Blot
TAOK1 is a serine/threonine-protein kinase involved in regulation of the p38-containing stress-responsive MAP kinase pathway and extracellular signal-regulated protein kinase (ERK) kinases (MEKs). The activation of and binding to MEK3 by TAOK1 implicates TAOK1 in the regulation of the p38-containing stress-responsive MAP kinase pathway. A microtubule affinity-regulating kinase, TAOK1 (also known as MARKK) is an important regulator of mitotic progression, required for both chromosome congression and checkpoint-induced anaphase delay. TAOK1 is involved in various processes such as p38/MAPK14 stress-activated MAPK cascase, DNA damage response, and regulations of cytoskeleton stability. It phosphorylates MAP2K3, MAP2K6, and MARK2. TAOK1 acts as an activator of the p38/MAPK cascade by mediating phosphorylation and subsequent activation of the upsteam MAP2K3 and MAP2K6 kinases. It is involved in G-protein couples receptor signaling to p38/MAPK14. In response to DNA damage, TAOK1, is involved in the G2/M transition DNA damage checkpoint by activating the p38/MAPK14 stress-activated MAPK cascade by phosphorylaton of MAP2K3 and MAP2K6. In addition, it acts as a regulator of cytoskeleton stability and a regulator of apoptosis.
Specifications
Specifications
| Antigen | TAOK1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2P66 |
| Concentration | 0.90 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | TAOK1 |
| Gene Accession No. | O88664, Q5F2E8, Q7L7X3 |
| Gene Alias | 2810468K05Rik; AU020252; D130018F14Rik; FLJ14314; h hTAOK1; H KIAA1361; hKFC-B; hTAOK1; KFC-B; Kiaa1361; Kinase from chicken homolog B; MAP3K16; MARK Kinase; Markk; microtubule affinity regulating kinase kinase; mKIAA1361; prostate-derived STE20-like kinase 2; prostate-derived sterile 20-like kinase 2; PSK2; PSK-2; serine/threonine protein kinase TAO1; serine/threonine protein kinase TAO1 homolog; serine/threonine-protein kinase TAO1; STE20-like kinase PSK2; tao; TAO kinase 1; TAO1; TAOK1; Thousand and one amino acid protein 1; thousand and one amino acid protein kinase 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?