missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAOK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26250-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It has similarity to rat protein which is thought to play a role in terminally differentiated neurons.Specifications
| TAOK1 | |
| Polyclonal | |
| Unconjugated | |
| TAOK1 | |
| hKFC B, KIAA1361, Kinase from chicken homolog B, MAP3K16, MARKK, PSK2, STE20 like kinase PSK2, TAO kinase 1, TAO1, TAOK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 216965, 57551 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5F2E8, Q7L7X3 | |
| TAOK1 | |
| TAOK1 Fusion Protein Ag22461 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction