missing translation for 'onlineSavingsMsg'
Learn More

TAO2 Antibody (2F4), Novus Biologicals™

Product Code. 18370569 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370569 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370569 Supplier Novus Biologicals Supplier No. H00009344M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAO2 Monoclonal antibody specifically detects TAO2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAO2
Applications Western Blot, ELISA, Immunocytochemistry, Proximity Ligation Assay
Classification Monoclonal
Clone 2F4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH51798
Gene Alias EC 2.7.11, KIAA0881PSK-1, Kinase from chicken homolog C, MAP3K17PSK1-BETA, Prostate-derived STE20-like kinase 1, PSK1EC 2.7.11.1, PSKhKFC-C, serine/threonine-protein kinase TAO2, TAO kinase 2, TAO1, TAO2prostate derived STE20-like kinase PSK, Thousand and one amino acid protein 2, thousand and one amino acid protein kinase
Host Species Mouse
Immunogen TAOK2 (AAH51798, 831 a.a. ~ 930 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ELQCRQYKRKMLLARHSLDQDLLREDLNKKQTQKDLECALLLRQHEATRELELRQLQAVQRTRAELTRLQHQTELGNQLEYNKRREQELRQKHAAQVRQQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9344
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.