missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAO Kinase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | TAO Kinase 1 |
|---|---|
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18421022
|
Novus Biologicals
NBP1-89864-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18256248
|
Novus Biologicals
NBP1-89864 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAO Kinase 1 Polyclonal specifically detects TAO Kinase 1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TAO Kinase 1 | |
| Polyclonal | |
| Rabbit | |
| GPCR, Protein Kinase | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| 57551 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LSPEAFSHSYPGASGWSHNPTGGPGPHWGHPMGGPPQAWGHPMQGGPQPWGHPSGPMQGVPRGSSMGVRNSPQALRRTASGGRTEQGMSRSTSVTSQISNGSH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.7.11, FLJ14314, hKFC-B, KIAA1361STE20-like kinase PSK2, Kinase from chicken homolog B, MAP3K16EC 2.7.11.1, MARKKmicrotubule affinity regulating kinase kinase, PSK2serine/threonine kinase TAO1, serine/threonine-protein kinase TAO1, TAO kinase 1, TAO1serine/threonine protein kinase TAO1 homolog, Thousand and one amino acid protein 1 | |
| TAOK1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title