missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) methoxide

Product Code. 11313497
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313497 1 g 1g
11323497 5 g 5g
11333497 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313497 Supplier Thermo Scientific Alfa Aesar Supplier No. 018839.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 865-35-0 | C5H15O5Ta | 336.118 g/mol

Tantalum(V) methoxide is used in the synthesis of tantalum hydride. Silica-supported alkoxo-tantalum complexes were used in the deperoxidation of cyclohexyl hydroperoxide into cyclohexanol and cyclohexanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-35-0
Molecular Formula C5H15O5Ta
Molecular Weight (g/mol) 336.118
MDL Number MFCD00058817
InChI Key QASMZJKUEABJNR-UHFFFAOYSA-N
PubChem CID 12787641
IUPAC Name methanolate;tantalum(5+)
SMILES C[O-].C[O-].C[O-].C[O-].C[O-].[Ta+5]

Specifications

Melting Point 50°C
Boiling Point 189°C (10 mmHg)
Linear Formula Ta(OCH3)5
Quantity 1 g
UN Number UN3181
Sensitivity Moisture Sensitive
Formula Weight 336.12
Physical Form Crystalline
Chemical Name or Material Tantalum(V) methoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.