missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane

Product Code. 11339227
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339227 100 mL 100mL
11349227 25 mL 25mL
11490241 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339227 Supplier Thermo Scientific Alfa Aesar Supplier No. 040038.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16761-83-4 | C15H40O5Ta | 481.428 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16761-83-4
Molecular Formula C15H40O5Ta
Molecular Weight (g/mol) 481.428
MDL Number MFCD00145347
InChI Key KWUQLGUXYUKOKE-UHFFFAOYSA-N
Synonym propan-2-olate,tantalum 5+
PubChem CID 131874823
IUPAC Name propan-2-ol;tantalum
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Ta]

Specifications

Odor Petrol-like
Linear Formula Ta[OCH(CH3)2]5
Quantity 100 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 476.39
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol/hexane
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tantalum(V) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.