missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)

Product Code. p-7046729
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11392867

Brand: Thermo Scientific Alfa Aesar L16033.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6074-84-6 | C10H25O5Ta | 406.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6074-84-6
Molecular Formula C10H25O5Ta
Molecular Weight (g/mol) 406.25
MDL Number MFCD00049785
InChI Key HSXKFDGTKKAEHL-UHFFFAOYSA-N
Synonym tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
PubChem CID 160806
IUPAC Name ethanolate;tantalum(5+)
SMILES CCO[Ta](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 145°C (0.1mmHg)
Melting Point 18°C to 19°C
Quantity 1 g
Linear Formula Ta(OCH2CH3)5
UN Number UN2924
Beilstein 3678999
Flash Point 30°C (86°F)
Formula Weight 406.25
Percent Purity 99.95%
Grade Optical
Refractive Index 1.488
Sensitivity Air and moisture sensitive
Density 1.566
Chemical Name or Material Tantalum(V) ethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.