missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) ethoxide, 99+%

Product Code. p-7045327
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11368345

Brand: Thermo Scientific Alfa Aesar L10288.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6074-84-6 | C10H25O5Ta | 406.25 g/mol

Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

Solubility
Soluble in organic solvents.

Notes
Moisture sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6074-84-6
Molecular Formula C10H25O5Ta
Molecular Weight (g/mol) 406.25
MDL Number MFCD00049785
InChI Key HSXKFDGTKKAEHL-UHFFFAOYSA-N
Synonym tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
PubChem CID 160806
SMILES CCO[Ta](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 145°C (0.1mmHg)
Melting Point 18°C to 19°C
Quantity 5 g
Linear Formula Ta(OCH2CH3)5
UN Number UN2924
Beilstein 3678999
Flash Point 30°C (86°F)
Solubility Information Soluble in organic solvents.
IUPAC Name ethanolate;tantalum(5+)
Formula Weight 406.25
Percent Purity ≥99%
Refractive Index 1.488
Sensitivity Air and moisture sensitive
Density 1.566
Chemical Name or Material Tantalum(V) ethoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tantalum(V) ethoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethanol, tantalum(5+) salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.