missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) chloride, Puratronic™, 99.99% (metals basis)

Product Code. 11367018
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367018 100 g 100g
11377018 5 g 5g
11387018 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367018 Supplier Thermo Scientific Alfa Aesar Supplier No. 014708.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7721-01-9 | Cl5Ta | 358.20 g/mol

Tantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 242°C
Melting Point 216°C
Color White to Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3260
Merck Index 14,9054
Solubility Information Soluble in ether,carbon tetrachloride,sulfuric acid and potassium hydroxide. Slightly soluble in ethanol.
Formula Weight 358.21
Percent Purity 99.99%
Grade Puratronic
Odor Acidic
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.68 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum(V) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.