Learn More
Alfa Aesar™ Tantalum(V) 2,2,2-trifluoroethoxide, 99.99+% (metals basis)
Brand: Alfa Aesar™ 039629.04
Code : NEW
Additional Details : CAS Number : 13053-54-8 Weight : 0.00200kg
Quantity | 2g |
---|
Chemical Identifiers
Tantalum(V) 2,2,2-trifluoroethoxide | |
Ta(OCH2CF3)5 | |
MFCD00145348 |
13053-54-8 | |
676.14 |
Specifications
Tantalum(V) 2,2,2-trifluoroethoxide | |
1.9g/mL | |
≥99.99% | |
Ta(OCH2CF3)5 | |
MFCD00145348 | |
>65° (>150°F) | |
Moisture Sensitive |
13053-54-8 | |
Liquid | |
(metals basis) | |
676.14 | |
115°C (0.1mmHg) | |
2g |