missing translation for 'onlineSavingsMsg'
Learn More

Tantalum tris(diethylamido)-tert-butylimide, 99.99% (metals basis)

Product Code. 11375707
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375707 1 g 1g
11365707 5 g 5g
11355707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375707 Supplier Thermo Scientific Alfa Aesar Supplier No. 043419.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Precursor for TaN | CAS: 169896-41-7 | C15H39N4Ta | 456.45 g/mol

Tantalum tris(diethylamido)-tert-butylimide is used as a precursor for tantalum nitride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169896-41-7
Molecular Formula C15H39N4Ta
Molecular Weight (g/mol) 456.45
MDL Number MFCD02684506
InChI Key NXXDVZOFTYVIKX-UHFFFAOYSA-N
Synonym tert-butylimino tris diethylamino tantalum, tantalum tris diethylamido-tert-butylimide, tbtdet, tris diethylamino tert-butylimino tantalum, t-butylimido tris diethylamino tantalum v, tris diethylamido tert-butylimido tantalum v, tris diethylamino tert-butylimino tantalum v, tert-butylimino bis diethylamino tantalio diethylamine, tantalum,tris n-ethylethanaminato 2-methyl-2-propanaminato 2--, t-4
PubChem CID 4100858
IUPAC Name tert-butyliminotantalum;diethylazanide
SMILES CCN(CC)[Ta](N(CC)CC)(N(CC)CC)=[N](C)(C)C

Specifications

Color Yellow
Boiling Point 90°C (0.1 mmHg)
Packaging Ampule
Quantity 1 g
UN Number UN3129
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 468.48
Percent Purity 99.99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tantalum tris (diethylamido)-tert-butylimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tantalum tris(diethylamido)-tert-butylimide
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.