missing translation for 'onlineSavingsMsg'
Learn More

Tantalum Thinfoil, 0.0127mm (0.0005in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11357857
Click to view available options
Quantity:
50 x 100 mm
50 x 500 mm
50 x 2000 mm
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.95% (metals basis)
Quantity 50 x 100 mm
Chemical Name or Material Tantalum Thinfoil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'