missing translation for 'onlineSavingsMsg'
Learn More

Tantalum carbide, 99.5% (metals basis)

Product Code. 11386508
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386508 250 g 250g
11396508 10 g 10g
11306518 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386508 Supplier Thermo Scientific Alfa Aesar Supplier No. 012144.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12070-06-3 | CTa | 192.96 g/mol

Tantalum carbides is used in  tungsten carbide. It is also used as a coating for steel moulds in the injection moulding of aluminum alloys. While providing a hard, wear resistant surface, it also provides a low friction mould surface. It is also used in the production of sharp instruments with extreme mechanical resistance and hardness, tool bits for cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-06-3
Molecular Formula CTa
Molecular Weight (g/mol) 192.96
MDL Number MFCD00011255
InChI Key DUMHRFXBHXIRTD-UHFFFAOYSA-N
IUPAC Name methanidylidynetantalumylium
SMILES [C-]#[Ta+]

Specifications

Boiling Point 5500°C
Melting Point 3880°C
Color Black to Gray
Physical Form Powder
Quantity 250 g
UN Number UN3178
Solubility Information Soluble in hydrofluoric acid and nitric acid. Partially soluble in water.
Formula Weight 192.96
Percent Purity 99.5%
Odor Odorless
Density 13.9 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum carbide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.