missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tanshinone IIA, MedChemExpress
Description
Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Specifications
Specifications
| Chemical Name or Material | Tanshinone IIA |
| CAS | 568-72-9 |
| Color | Red |
| Purity Grade Notes | Research |
| Molecular Formula | C19H18O3 |
| Quantity | 10 mg |
| Synonym | Dan Shen ketone |
| Solubility Information | DMSO : 100 mg/mL (339.74 mM; Need ultrasonic) |
| SMILES | O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC=C4C)=O |
| Molecular Weight (g/mol) | 294.34 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?