missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid, 95%, Thermo Scientific Chemicals

Product Code. 10702441
Click to view available options
Quantity:
5 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

Melting Point 218.0°C
Color White to Yellow
Assay Percent Range 95%
Merck Index 15, 9184
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: soluble in glycerol, ethanol and acetone;, practically insoluble in benzene, chcl3, diethyl e, ther, petroleum ether, cs2, ccl4, hexane;, slightly soluble in anhydrous ethanol.
Formula Weight 1701.23
Percent Purity 95%
Density 198g/mL
Quantity 1 kg
Flash Point 198°C
Infrared Spectrum Authentic
Loss on Drying 12.0% max.
Packaging Plastic bottle
Physical Form Beads
Chemical Name or Material Tannic acid
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'