missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid

Product Code. 11491781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491781 500 g 500g
11401791 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491781 Supplier MP Biomedicals Supplier No. 0219027590
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H18Cl2N2
Molecular Weight (g/mol) 237.168
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl

Specifications

Color Yellow
Quantity 500 g
Physical Form Powder
Chemical Name or Material Tannic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.