missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tankyrase binding protein 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £571.00
Specifications
| Antigen | Tankyrase binding protein 1 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18422872
|
Novus Biologicals
NBP1-89826-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18431792
|
Novus Biologicals
NBP1-89826 |
0.1 mL |
£571.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tankyrase binding protein 1 Polyclonal antibody specifically detects Tankyrase binding protein 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Tankyrase binding protein 1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KIAA1741TAB182FLJ45975,182 kDa tankyrase-1-binding protein, tankyrase 1 binding protein 1, 182kDa, tankyrase 1-binding protein of 182 kDa | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DWTPDLGLRNMAPGAVCSPGESKELGVGQMDWGNNLGLRDLEVTCDPDSGGSQGLRGCGVGQMDWTQDLAPQNVELFGAPSEARE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 85456 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title