missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tankyrase 2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518848
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Poly-ADP-ribosyltransferase involved in various processes such as Wnt signaling pathway, telomere length and vesicle trafficking (PubMed:11739745, PubMed:11802774, PubMed:19759537, PubMed:21478859, PubMed:23622245, PubMed:25043379). Acts as an activator of the Wnt signaling pathway by mediating poly-ADP-ribosylation of AXIN1 and AXIN2, 2 key components of the beta-catenin destruction complex: poly-ADP-ribosylated target proteins are recognized by RNF146, which mediates their ubiquitination and subsequent degradation (PubMed:19759537, PubMed:21478859). Also mediates poly-ADP-ribosylation of BLZF1 and CASC3, followed by recruitment of RNF146 and subsequent ubiquitination (PubMed:21478859). Mediates poly-ADP-ribosylation of TERF1, thereby contributing to the regulation of telomere length (PubMed:11739745). Stimulates 26S proteasome activity (PubMed:23622245). [UniProt]Specifications
Tankyrase 2 | |
Polyclonal | |
Unconjugated | |
TNKS2 | |
5430432P15Rik; AA517131; ADP-ribosyltransferase diphtheria toxin-like 6; AI662480; ARTD6; PARP5B; PARP-5b; PARP5C; PARP-5c; pART6; Poly [ADP-ribose] polymerase 5B; Poly [ADP-ribose] polymerase tankyrase-2; Protein poly-ADP-ribosyltransferase tankyrase-2; TANK2; tankyrase 2; tankyrase II; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2; tankyrase-2; Tankyrase-like protein; tankyrase-related protein; TNKL; tnks 2; TNKS2; TNKS-2; TRF1-interacting ankyrin-related ADP-ribose polymerase 2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
80351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H2K2 | |
TNKS2 | |
Synthetic peptide sequence (HRRKEVSEENHNH) corresponding to the internal amino acids of TNKS2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |