missing translation for 'onlineSavingsMsg'
Learn More

Tankyrase 1 Antibody [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30733572 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733572 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733572 Supplier Novus Biologicals Supplier No. AF7116I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Tankyrase 1 Polyclonal antibody specifically detects Tankyrase 1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tankyrase 1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias EC 2.4.2.30, PARP-5a, PARP5APARPL, pART5, Poly [ADP-ribose] polymerase 5A, Tankyrase I, tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase, tankyrase-1, TIN1TANK1, TINF1TNKS-1, TNKS1TRF1-interacting ankyrin-related ADP-ribose polymerase
Host Species Sheep
Immunogen E. coli-derived recombinant human Tankyrase 1, Ala943-Gly1039, Accession # O95271
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 8658
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.