missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TANK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56144SS
This item is not returnable.
View return policy
Description
TANK Polyclonal specifically detects TANK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TANK | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| ITRAF, I-TRAFTRAF2, TRAF family member-associated NF-kappa-B activator, TRAF family member-associated NFKB activator, TRAF-interacting protein | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TANK | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| TANK | |
| A synthetic peptide corresponding to amino acids 346-367 (DPSDAPFPSLDSPGKAIRGPQQ) of human ITRAF was used as immunogen, GeneBank no. AAC50681.1. | |
| 0.025 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 10010 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction