missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TANK Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95201-0.02ml
This item is not returnable.
View return policy
Description
TANK Polyclonal antibody specifically detects TANK in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TANK | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ITRAF, I-TRAFTRAF2, TRAF family member-associated NF-kappa-B activator, TRAF family member-associated NFKB activator, TRAF-interacting protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-119 of human TANK (NP_597841.1). MDKNIGEQLNKAYEAFRQACMDRDSAVKELQQKTENYEQRIREQQEQLSLQQTIIDKLKSQLLLVNSTQDNNYGCVPLLEDSETRKNNLTLDQPQDKVISGIAREKLPKVDIASAESSI | |
| 0.02 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 10010 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction