missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango™ GPR120S-bla U2OS Cells

Product Code. 10105654
Change view
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10105654 8 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10105654 Supplier Thermo Scientific™ Supplier No. K1845

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tango™ GPR120 (short)-bla U2OS cells contain the human GPR120 (short) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

Tango™ GPR120 (short)-bla U2OS cells contain the human GPR120 (short) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR120 (short)-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of GW9508 (Figure 1). In addition, Tango™ GPR120 (short)-bla U2OS cells have been tested for assay performance under variable conditions.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_181745.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D, and other materials are required separately; please refer to the protocol.

Includes:
• Tango™ GPR120S-bla U2OS cells
• Tango™ GPR120S-bla U2OS cell Assay Protocol
• Certificate of Analysis

The Tango™ GPR120S-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Native Tail
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol GPR120S
Human Protein Class G-Protein Coupled Receptor (GPCR)
Includes GPR120S-bla U2OS cells, GPR120S-bla U2OS cell Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 8 x 106 Cells
Readout End Point
Target Entry GPR120S
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.