Learn More
Thermo Scientific™ Tango™ CNR1-bla U2OS Cells
Description
The Tango™ CNR1-bla U2OS cells contain the human Cannabinoid Receptor 1 (CNR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ CNR1 cell line has been functionally validated for Z'-factor and EC50 concentrations of CP 55,940. The CNR1 cells have been tested for assay performance under variable assay conditions. These data are available in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
Specifications
| Accession Number | NM_016083 |
| Assay Entry | Cell-based beta-arrestin recruitment |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ CNR1-bla U2OS cells • Tango™ CNR1-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ CNR1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Cytoplasmic Tail | Tail from AVPR2 GPCR |
| Detection Method | Fluorescence |
| For Use With (Application) | GPCRs |
| Gene Symbol | CNR1 |
| Includes | CNR1-bla U2OS cells, CNR1-bla U2OS Assay Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.