missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango™ CCR6-bla U2OS Cells

Product Code. 10054004
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10054004 2 x 106 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10054004 Supplier Thermo Scientific™ Supplier No. K1761

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Tango™ CCR6-bla U2OS cells contain the human Chemokine (C-C motif) Receptor 6 (CCR6) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ CCR6-bla U2OS cells cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ CCR6-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_004367.3
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage K1761 Tango™ CCR6-bla U2OS cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• Tango™ CCR6-bla U2OS cells
• Tango™ CCR6-bla U2OS Assay Protocol
• Certificate of Analysis

Storage:
The Tango™ CCR6-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Native Tail
Detection Method Fluorescence
Gene Symbol CCR6
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry CCR6
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.