missing translation for 'onlineSavingsMsg'
Learn More

Tangeretin, MedChemExpress

Product Code. 16458996
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458996 100 mg 100mg
16424767 50 mg 50mg
16408878 10 mg 10mg
16401318 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16458996 Supplier Medchem Express Supplier No. HYN0133

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tangeretin (Tangeritin), a flavonoid from citrus fruit peels, has been proven to play an important role in anti-inflammatory responses and neuroprotective effects in several disease models, and is a Notch-1 inhibitor.

Tangeretin (Tangeritin), a flavonoid from citrus fruit peels, has been proven to play an important role in anti-inflammatory responses and neuroprotective effects in several disease models, and is a Notch-1 inhibitor.

Specifications

Chemical Name or Material Tangeretin
CAS 481-53-8
Color White
Purity Grade Notes Research
Molecular Formula C20H20O7
Quantity 100 mg
Synonym Tangeritin NSC53909 NSC618905
Solubility Information DMSO : 25 mg/mL (67.14 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C=C2)OC3=C(OC)C(OC)=C(OC)C(OC)=C13
Molecular Weight (g/mol) 372.37
Formula Weight 372.37
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.