missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tangential Flow Filtration (TFF)-EV Concentrator

Product Code. 18628161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Units
5 Units
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18628161 1 Units Each
18650831 5 Units Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18628161 Supplier Novus Biologicals™ Supplier No. NBP3117621Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Content And Storage Store at room temperature.
Quantity 1 Units
Sample Type Diluted matrices (urine and cell culture supernatant)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.