missing translation for 'onlineSavingsMsg'
Learn More

Tanespimycin, MedChemExpress

Product Code. p-200096460
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
This item is not returnable. View return policy

Product Code. 16482836

Brand: Medchem Express HY10211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tanespimycin (17-AAG) is a potent HSP90 inhibitor with an IC50 of 5 nM, having a 100-fold higher binding affinity for tumour cell derived HSP90 than normal cell derived HSP90. Tanespimycin depletes cellular STK38/NDR1 and reduces STK38 kinase activity. Tanespimycin also downregulates the stk38 gene expression.

Tanespimycin (17-AAG) is a potent HSP90 inhibitor with an IC50 of 5 nM, having a 100-fold higher binding affinity for tumour cell derived HSP90 than normal cell derived HSP90. Tanespimycin depletes cellular STK38/NDR1 and reduces STK38 kinase activity. Tanespimycin also downregulates the stk38 gene expression.

Specifications

Chemical Name or Material Tanespimycin
Physical Form Powder
CAS 75747-14-7
Color Pale Purple
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C31H43N3O8
For Use With (Application) Cancer-Kinase/protease
Synonym 17-AAG NSC 330507 CP 127374
Solubility Information DMSO : 50 mg/mL (85.37 mM; Need ultrasonic)
SMILES O=C(C(NC(/C(C)=C/C=C\[C@H](OC)[C@H](/C(C)=C/[C@@H]([C@H]([C@H](C[C@@H](C1)C)OC)O)C)OC(N)=O)=O)=CC2=O)C1=C2NCC=C
Molecular Weight (g/mol) 585.69
Formula Weight 585.69
Percent Purity 97.41%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.