missing translation for 'onlineSavingsMsg'
Learn More

Tanespimycin, MedChemExpress

Product Code. 16482836
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482836 25 mg 25mg
16464786 100 mg 100mg
16423718 10 mg 10mg
16410956 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482836 Supplier Medchem Express Supplier No. HY10211
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tanespimycin (17-AAG) is a potent HSP90 inhibitor with an IC50 of 5 nM, having a 100-fold higher binding affinity for tumour cell derived HSP90 than normal cell derived HSP90. Tanespimycin depletes cellular STK38/NDR1 and reduces STK38 kinase activity. Tanespimycin also downregulates the stk38 gene expression.

Tanespimycin (17-AAG) is a potent HSP90 inhibitor with an IC50 of 5 nM, having a 100-fold higher binding affinity for tumour cell derived HSP90 than normal cell derived HSP90. Tanespimycin depletes cellular STK38/NDR1 and reduces STK38 kinase activity. Tanespimycin also downregulates the stk38 gene expression.

Specifications

Chemical Name or Material Tanespimycin
Physical Form Powder
CAS 75747-14-7
Color Pale Purple
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C31H43N3O8
For Use With (Application) Cancer-Kinase/protease
Synonym 17-AAG NSC 330507 CP 127374
Solubility Information DMSO : 50 mg/mL (85.37 mM; Need ultrasonic)
SMILES O=C(C(NC(/C(C)=C/C=C\[C@H](OC)[C@H](/C(C)=C/[C@@H]([C@H]([C@H](C[C@@H](C1)C)OC)O)C)OC(N)=O)=O)=CC2=O)C1=C2NCC=C
Molecular Weight (g/mol) 585.69
Formula Weight 585.69
Percent Purity 97.41%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.