missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tandospirone citrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
Tandospirone citrate is a potent and selective 5-HT1A receptor partial agonist (Ki = 27 nM) that displays selectivity over SR-2, SR-1C, α1, α2, D1 and D2 receptors (Ki values ranging from 1300-41000 nM).
Specifications
Specifications
| Chemical Name or Material | Tandospirone citrate |
| CAS | 112457-95-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H37N5O9 |
| Quantity | 25 mg |
| Synonym | SM-3997 citrate |
| Solubility Information | H2O : 10.4 mg/mL (18.07 mM; Need ultrasonic) |
| SMILES | O=C1N(CCCCN2CCN(C3=NC=CC=N3)CC2)C([C@@]4([H])[C@](C5)([H])CC[C@]5([H])[C@@]14[H])=O.O=C(CC(C(O)=O)(O)CC(O)=O)O |
| Molecular Weight (g/mol) | 575.61 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction