missing translation for 'onlineSavingsMsg'
Learn More

Tandospirone citrate, MedChemExpress

Product Code. 16418338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16418338

Brand: Medchem Express HYB0061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tandospirone citrate is a potent and selective 5-HT1A receptor partial agonist (Ki = 27 nM) that displays selectivity over SR-2, SR-1C, α1, α2, D1 and D2 receptors (Ki values ranging from 1300-41000 nM).

Tandospirone citrate is a potent and selective 5-HT1A receptor partial agonist (Ki = 27 nM) that displays selectivity over SR-2, SR-1C, α1, α2, D1 and D2 receptors (Ki values ranging from 1300-41000 nM).

Specifications

Chemical Name or Material Tandospirone citrate
CAS 112457-95-1
Color White
Purity Grade Notes Research
Molecular Formula C27H37N5O9
Quantity 10 mg
Synonym SM-3997 citrate
Solubility Information H2O : 10.4 mg/mL (18.07 mM; Need ultrasonic)
SMILES O=C1N(CCCCN2CCN(C3=NC=CC=N3)CC2)C([C@@]4([H])[C@](C5)([H])CC[C@]5([H])[C@@]14[H])=O.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 575.61
Formula Weight 575.61
Percent Purity 98.79%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.