missing translation for 'onlineSavingsMsg'
Learn More

TANC1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18409442 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409442 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409442 Supplier Novus Biologicals Supplier No. NBP152630JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TANC1 Polyclonal antibody specifically detects TANC1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TANC1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ13360, FLJ41166, KIAA1728, protein TANC1, rolling pebbles homolog B, ROLSB, TANC, tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1, tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1, TPR domain, ankyrin-repeat and coiled-coil-containing
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Tetratricopeptide Repeat, Ankyrin Repeat and Coiled-Coil Containing 1 using the numbering given in entry NP_203752.2 (GeneID 85461).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85461
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.