missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAMRA Monoclonal Antibody (11H10)
GREENER_CHOICE

Product Code. 12807772
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807772 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12807772 Supplier Invitrogen™ Supplier No. MA1042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-042 has been successfully used in fluorescence quenching applications.

The red-fluorescent tetramethylrhodamine (TAMRA) azide can be used as a reactive-azido group (Staudinger reagents) that facilitates multiplex labeling when used with phosphine- or alkyne-derivatized tags. In addition, TAMRA fluorophore has been used to label and detect active serine hydrolases in samples by fluorescent gel imaging, capillary electrophoresis or mass spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAMRA
Applications Fluorescent Quenching
Classification Monoclonal
Clone 11H10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Alias TAMRA
Host Species Mouse
Immunogen TAMRA molecule.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.