missing translation for 'onlineSavingsMsg'
Learn More

TAMRA Antibody (5G5), Novus Biologicals™

Mouse Monoclonal Antibody

£174.00

Specifications

Antigen TAMRA
Clone 5G5
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Immunoprecipitation 10 ug
Applications Western Blot, Immunoprecipitation
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18773394
View Documents
Novus Biologicals
NBP2-22472-0.025mg
0.025 mg
£174.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TAMRA Monoclonal specifically detects TAMRA in Non-species specific samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

TAMRA
1 mg/mL
Western Blot, Immunoprecipitation
Unconjugated
Mouse
Non-species specific
TAMRA molecule
Primary
Store at -20°C. Avoid freeze-thaw cycles.
5G5
Western Blot 1:1000, Immunoprecipitation 10 ug
Monoclonal
Purified
RUO
BP-25606, F07-0040, TAMRA Azide, TAMRA Azide, isomer 5, tetramethylrhodamine azide
IgG1
Protein G purified
This antibody reacts with Chemical.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.