missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Tamper Evident Screw Caps

Product Code. 17466923 Shop All Bürkle Products
Click to view available options
Thread Size (Metric):
DIN 45
Unit Size:
Each

Product Code. 17466923

Brand: Buerkle™ 14279107

Only null left
Add to basket
Edge
Add to basket

Buerkle™ Tamper Evident Screw Caps with DIN50/ DIN45 thread size is suitable for use with jerrycans.

  • Tamper evident
  • DIN45/DIN50 thread size
  • For use with jerrycans
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description Original screw cap
Material HDPE
Quantity 1
Thread Size (Metric) DIN 45
Neck Finish 42 mm
Product Type Tamper-evident Screw Cap
For Use With (Equipment) Jerrycans
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.