missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen N-Oxide, TRC

Product Code. 30409979
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409979 1 mg
30414697 50 mg
30416642 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409979 Supplier Toronto Research Chemicals Supplier No. TRCT0060951MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75504-34-6
Molecular Formula C26 H29 N O2
Molecular Weight (g/mol) 387.51
Synonym Tamoxifen N-Oxide
IUPAC Name 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine oxide
SMILES CC\C(=C(/c1ccccc1)\c2ccc(OCC[N+](C)(C)[O-])cc2)\c3ccccc3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H29NO2/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)29-20-19-27(2,3)28/h5-18H,4,19-20H2,1-3H3/b26-25-
Formula Weight 387.2198
Chemical Name or Material Tamoxifen N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.