missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, MP Biomedicals

Product Code. 11455161 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455161 1 g 1g
15712262 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455161 Supplier MP Biomedicals Supplier No. 0215673891
  • ONLINE EXCLUSIVE PRICE
    £407.00
    £381.15 / 1g
    Save £25.85
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Tamoxifen, White Powder

  • Tamoxifen has been used to facilitate the recombination of ect2flox allele in mouse organs
  • It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10540-29-1
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
MDL Number MFCD00010454
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
PubChem CID 2733526
ChEBI CHEBI:41774
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C to 100°C
Color White
Linear Formula C26H29NO
Quantity 1 g
Synonym tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
Solubility Information Practically insoluble in water (< 0.01% at 20°C); soluble in methanol, ethanol, 2-propanol, propylene glycol, chloroform (50mg/mL - clear, colorless to faint yellow solution) or DMSO.
IUPAC Name 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine
Formula Weight 371.5
Physical Form Powder
Chemical Name or Material TAMOXIFEN
compliance-icons
Product Identifier
  • Tamoxifen, Free Base
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P263-Avoid contact during pregnancy/while nursing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.