missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen, MedChemExpress

Product Code. 16491379
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491379 1 g 1g
16431706 500 mg 500mg
16488497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491379 Supplier Medchem Express Supplier No. HY13757A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM) which blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells.

Tamoxifen is a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity. Tamoxifen also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.1 μM and 1.8 μM, respectively. Tamoxifen activates autophagy and induces apoptosis. Tamoxifen also can induce gene knockout of CreER(T2) transgenic mouse.

Specifications

Chemical Name or Material Tamoxifen
CAS 10540-29-1
Color White
Purity Grade Notes Research
Molecular Formula C26H29NO
Quantity 1 g
Synonym ICI 47699 (Z)-Tamoxifen trans-Tamoxifen
Solubility Information DMSO : 50 mg/mL (134.59 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (134.59 mM; Need ultrasonic)
SMILES CC/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(C)C)C=C2)\C3=CC=CC=C3
Molecular Weight (g/mol) 371.51
Formula Weight 371.51
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.