missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen citrate

Product Code. 15423639
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423639 5 g 5g
15413639 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423639 Supplier Thermo Scientific Alfa Aesar Supplier No. J60955.06
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of protein kinase C and induces apoptosis in cancer cells | CAS: 54965-24-1 | C32H37NO8 | 563.65 g/mol

Tamoxifen citrate is used as an antiestrogen with potent anti-cancer and PKC inhibitory activities. Tamoxifen Citrate functions as an estrogen receptor antagonist in breast tissue. Tamoxifen Citrate is an agonist towards this receptor in tissues of the endometrium. Tamoxifen Citrate inhibits DNA synthesis and transcription by recruiting co-repressors, which stop estrogen from interacting with genes. When combined with PAX2, Tamoxifen Citrate can inhibit the proliferation of ERBB2. It is also a high affinity activator of the GPR30 receptor. Tamoxifen Citrate is an activator of Estrogen Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tamoxifen citrate is used as an antiestrogen with potent anti-cancer and PKC inhibitory activities. Tamoxifen Citrate functions as an estrogen receptor antagonist in breast tissue. Tamoxifen Citrate is an agonist towards this receptor in tissues of the endometrium. Tamoxifen Citrate inhibits DNA synthesis and transcription by recruiting co-repressors, which stop estrogen from interacting with genes. When combined with PAX2, Tamoxifen Citrate can inhibit the proliferation of ERBB2. It is also a high affinity activator of the GPR30 receptor. Tamoxifen Citrate is an activator of Estrogen Receptor.

Solubility
Soluble in methanol (50 mg/ml with heat) or ethanol (10 mg/ml with sonication); very slightly soluble in water (0.3 mg/L @ 20°C; the pH is approximately 3.0-3.5), 0.02 N HCl (0.2 mg/ml @ 37°C)4, acetone ,chloroform,and DMSO(100mM)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54965-24-1
Molecular Formula C32H37NO8
Molecular Weight (g/mol) 563.65
MDL Number MFCD00058321
InChI Key FQZYTYWMLGAPFJ-OQKDUQJOSA-N
Synonym tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen
PubChem CID 2733525
ChEBI CHEBI:9397
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9048
Solubility Information Soluble in methanol (50mg/ml with heat) or ethanol (10mg/ml with sonication); very slightly soluble in water (0.3mg/L @ 20°C; the pH is approximately 3.0-3.5),0.02 N HCl (0.2mg/ml @ 37°C)4,acetone ,chloroform,and DMSO(100mM)
Formula Weight 563.65
Physical Form Powder
Chemical Name or Material Tamoxifen citrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tamoxifen citrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tamoxifen citrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.